Order Entry
Sweden
ContactUsLinkComponent
Anti-TH Rabbit Polyclonal Antibody
Anti-TH Rabbit Polyclonal Antibody
Catalog # CAYM10009414-1
Supplier:  Cayman Chemical
Anti-TH Rabbit Polyclonal Antibody
Catalog # CAYM10009414-1
Supplier:  Cayman Chemical

Specifications

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    TH
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Reaktivitet:
    Multiple
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Modifiering:
    phospho-Ser40
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    phosphopeptide corresponding to amino acid residues surrounding phospho-Ser40 of rat TH
  • Rening:
    purified by affinity chromatography
  • Storlek:
    1 Ea
  • Förp.:
    1

Specifications

About this item

Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mammal, non-Mammal species