Order Entry
Sweden
ContactUsLinkComponent
Anti-TPH Rabbit Polyclonal Antibody
Anti-TPH Rabbit Polyclonal Antibody
Katalog # CAYM10009397-1
Leverantör:  Cayman Chemical
Anti-TPH Rabbit Polyclonal Antibody
Katalog # CAYM10009397-1
Leverantör:  Cayman Chemical

Specifikationer

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    TPH
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • Reaktivitet:
    Rabbit
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Modifiering:
    phospho-Ser58
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    phosphopeptide corresponding to amino acid residues surrounding phospho-Ser58 of TPH
  • Rening:
    purified by affinity chromatography
  • Storlek:
    1 Ea
  • Förp.:
    1

Specifikationer

Om denna produkt

Type: Primary
Antigen: TPH
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rabbit