Anti-LCAT Rabbit Polyclonal Antibody
Catalog # CAYM10009323-500
Leverantör: Cayman Chemical
Specifikationer
- Antikroppstyp:Primär
- Antigensymbol:LCAT
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Isotyp:IgG
- Reaktivitet:Pig,Human,Bovine,Mouse
- Western blot:Yes
- Korsad adsorption:No
- Aminosyretal:132 to 143
- Sequence:SVEYLDSSKLAG
- Förvaringstemperatur:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human LCAT protein amino acids 132-143
- Rening:purified by affinity chromatography
- Storlek:500 µl
- Förp.:500 µl
Specifikationer
Om denna produkt
Type: Primary
Antigen: LCAT
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Pig