Anti-IgG (H+L) Rabbit Polyclonal Antibody (DyLight® 800)
ROCK610-445-002-05
- Antikroppstyp:Sekundär
- Antigensymbol:IgG (H+L)
- Klonalitet:Polyclonal
- Konjugation:DyLight® 800
- Värd:Rabbit
- ImmunoFluorescens:Yes
- Reaktivitet:Mouse
- Western blot:Yes
- Storlek:500 µg
- Förp.:500 µG
Type: Secondary
Antigen: IgG(Mouse)-HL
Clonality: Polyclonal
Clone:
Conjugation: DyLight800
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse