Anti-CPB1 Rabbit Polyclonal Antibody
Catalog # ROCK200-401-049S
Supplier: Rockland Immunochemicals
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:carboxypeptidase B1 (tissue)
- Antigensymbol:CPB1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- Reaktivitet:Pig,Bovine
- Western blot:Yes
- Korsad adsorption:No
- Gen-ID:397341
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Förvaringstemperatur:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Carboxypeptidase B [Porcine Pancreas]
- Rening:purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Storlek:25 µl
- Förp.:25 µl
Specifications
About this item
Anti-Carboxypeptidase B antibody is suitable for ELISA, Western Blot and Immunohistochemistry applications. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500- 1:2000; Immunochemistry: 1:250 - 1:1000; Other Dilution: User Optimized
Type: Primary
Antigen: CPB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig, Bovine