Anti-SERPINA1 Goat Polyclonal Antibody
Katalog # ROCK200-101-147S
Leverantör: Rockland Immunochemicals
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1
- Antigensymbol:SERPINA1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Goat
- Reaktivitet:Human
- Western blot:Yes
- Korsad adsorption:No
- Gen-ID:5265
- Antigen synonymer:alpha1AT|PRO2275|AAT|PI|A1A|PI1|A1AT
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Förvaringstemperatur:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:a1-Anti-Trypsin [Human Plasma]
- Rening:purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Storlek:25 µl
- Förp.:25 µl
Specifikationer
Om denna produkt
ALPHA-1-ANTI-TRYPSIN Antibody has been assayed against 1.0 ug of a1-Anti-Trypsin [Human Plasma] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG [H&L] (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Other Dilution: User Optimized
Type: Primary
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human