Anti-IL4 Rabbit Polyclonal Antibody (Biotin)
Catalog # ROCK209-406-B93
Supplier: Rockland Immunochemicals
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:interleukin 4
- Antigensymbol:IL4
- Klonalitet:Polyclonal
- Konjugation:Biotin
- ELISA:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Human
- Western blot:Yes
- Epitop:Full length
- Korsad adsorption:No
- Mått:Antibody fraction
- Form:Lyophilized
- Gen-ID:3565
- Antigen synonymer:BCGF-1|IL-4|BCGF1|BSF1|BSF-1
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
- Förvaringstemperatur:–20 °C
- Shipping temperature:Ambient
- Immunogen:This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant human IL-4 Protein.
- Rening:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
This purified antibody has been tested for use in ELISA and western blotting. By western blot a band approximately 15 kDa in size corresponding to human IL-4 Protein is expected in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: IL4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human