Anti-TLR7 Rabbit Polyclonal Antibody
Catalog # ROCK200-401-A64
Supplier: Rockland Immunochemicals
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Toll-like Receptor 7
- Antigensymbol:TLR7
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Human,Mouse
- Western blot:Yes
- Epitop:Internal
- Korsad adsorption:No
- Mått:Antibody fraction
- Form:Liquid (sterile filtered)
- Gen-ID:170743
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Förvaringstemperatur:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:TLR7 Antibody was produced from whole rabbit serum prepared by repeated immunizations with a peptide corresponding to an internal region of mouse TLR7.
- Rening:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
TLR7 Antibody has been tested for use in ELISA, immunochemistry and western blotting. Expect a band at ~121 kDa in size corresponding to TLR7 by western blotting in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1 -2 µg/ml; Immunochemistry: 2 µg/ml;
Type: Primary
Antigen: TLR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse