Anti-3CL-PRO Rabbit Polyclonal Antibody
Catalog # ROCK200-401-A51
Supplier: Rockland Immunochemicals
Specifications
- Antikroppstyp:Primär
- Antigensymbol:3CL-PRO
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:SARS virus
- Western blot:Yes
- Epitop:Full length
- Korsad adsorption:No
- Mått:Antibody fraction
- Form:Lyophilized
- Gen-ID:1489680
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Förvaringstemperatur:–20 °C
- Shipping temperature:Ambient
- Immunogen:This Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a purified recombinant Protein corresponding to full length SARS Coronavirus 3CL Protease. Lifesensors Inc. (www.lifesensors.com) prepared the 3
- Rening:Protein A purified
- Storlek:500 µg
- Förp.:500 µG
Specifications
About this item
This Protein A purified antibody has been tested for use in ELISA and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 34 kDa in size corresponding to SARS 3CL Protease by western blottiing.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:10000
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: 3CL-PRO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: SARS