Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Katalog # ROCK610-1519-0500
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Katalog # ROCK610-1519-0500

Specifikationer

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgG (H+L)
  • Klonalitet:
    Polyclonal
  • Konjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Värd:
    Goat
  • ImmunKemi:
    Yes
  • Reaktivitet:
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Storage buffer:
    0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
  • Förvaringstemperatur:
    +4 °C
  • Shipping temperature:
    Wet Ice
  • Immunogen:
    Mouse IgG whole molecule
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    500 µg
  • Förp.:
    500 µG

Specifikationer

Om denna produkt

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500 - 1:4000; Immunochemistry: 1:500 - 1:2000

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse