Anti-HSP90AB1 Mouse Monoclonal Antibody (Unconjugated) [clone: D7A]
Katalog # ROCK200-301-F68
Leverantör: Rockland Immunochemicals
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Heat shock protein 90kDa alpha (cytosolic), class B member 1
- Antigensymbol:HSP90AB1
- Klonalitet:Monoclonal
- Klon:D7A
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Mouse
- ImmunKemi:Yes
- Immunutfällning:Yes
- Isotyp:IgG
- Reaktivitet:Pig,Rabbit,Chicken,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Epitop:Full length
- Korsad adsorption:No
- Mått:Antibody fraction
- Form:Liquid (sterile filtered)
- Gen-ID:3326
- Antigen synonymer:HSP84|D6S182|HSPCB|HSPC2|HSP90B
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Förvaringstemperatur:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Hsp90 Antibody was produced in mice by repeated immunizations raised against full length Protein Hsp90 purified from chicken brain.
- Rening:Protein G purified
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Anti-Hsp90 Antibody is suitable for WB, IP, ELISA, and IHC. Expect a band approximately 90kD Proteins corresponding to the molecular mass of Hsp90 on SDS Page immunoblots. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:200; Western Blot: 1:500; Immunoprecipitation: 5µg/20µl
Type: Primary
Antigen: HSP90AB1
Clonality: Monoclonal
Clone: D7A
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG
Reactivity: Bovine, Chicken, Human, Mouse, Pig, Rat, Rabbit