Anti-STAT6 Rabbit Polyclonal Antibody
Katalog # ROCK600-401-C67
Leverantör: Rockland Immunochemicals
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:signal transducer and activator of transcription 6, interleukin-4 induced
- Antigensymbol:STAT6
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- Isotyp:IgG
- Reaktivitet:Human
- Western blot:Yes
- Korsad adsorption:No
- Mått:Antibody fraction
- Form:Liquid (sterile filtered)
- Gen-ID:6778
- Antigen synonymer:STAT6C|IL-4-STAT|D12S1644|STAT6B
- Modifiering:phospho-Tyr641
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Förvaringstemperatur:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:STAT6 phospho Y641 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding Y641 of human STAT6 Protein.
- Rening:affinity-purified
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
This affinity purified antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 94.1kDa in size corresponding to STAT6 pY641 Protein by western blotting.
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1:1000;
Type: Primary
Antigen: STAT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human