Anti-HUWE1 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-C03
Supplier: Rockland Immunochemicals
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
- Antigensymbol:HUWE1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Korsad adsorption:No
- Mått:Antibody fraction
- Form:Liquid (sterile filtered)
- Gen-ID:10075
- Antigen synonymer:URE-B1|HSPC272|MULE|ARF-BP1|Ib772|LASU1|HECTH9|UREB1
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Förvaringstemperatur:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:ARF-BP1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the carboxy terminus of human ARF-BP1 Protein.
- Rening:affinity-purified
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
ARF-BP1 antibody has been tested in ELISA, WB, and ICC. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~481 kDa in size corresponding to ARF-BP1 by Western blotting in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:10000 - 1:40000; Immunofluorescence: 5.0 µg/ml; Western Blot: 1.0 -2.0 µg/ml;
Type: Primary
Antigen: HUWE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human