Anti-IgG1/IgG2a/IgG2b/IgG3 gamma Rabbit Polyclonal Antibody (ATTO 488)
Catalog # ROCK610-452-C46S
Supplier: Rockland Immunochemicals
Specifications
- Antikroppstyp:Sekundär
- Antigensymbol:IgG1/IgG2a/IgG2b/IgG3 gamma
- Klonalitet:Polyclonal
- Konjugation:ATTO 488
- Värd:Rabbit
- ImmunoFluorescens:Yes
- Reaktivitet:Mouse
- Western blot:Yes
- ChIP:Yes
- Tested applications:Fluoresence ELISA, Fluorescence Resonance Energy Transfer, Stimulated Emission Depletion Microscopy
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse