Order Entry
Sweden
ContactUsLinkComponent
Anti-IgM µ Rabbit Polyclonal Antibody
Anti-IgM µ Rabbit Polyclonal Antibody
Catalog # ROCK610-4107-0500
Anti-IgM µ Rabbit Polyclonal Antibody
Catalog # ROCK610-4107-0500

Specifications

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgM µ
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Reaktivitet:
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    Wet Ice
  • Immunogen:
    Anti-Mouse IgM mu heavy chain antibody was produced by repeated immunization with Mouse IgM mu heavy chain in rabbit.
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    500 µg
  • Förp.:
    500 µG

Specifications

About this item

Anti-Mouse IgM mu heavy chain antibody is suitable for highly specific immunological methods requiring extremely low background levels, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:500 - 1:2500; Other Dilution: User Optimized

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Rabbit
Isotype:
Reactivity: Mouse