Anti-SYT1 Rabbit Polyclonal Antibody
Katalog # ROCK612-401-E37
Leverantör: Rockland Immunochemicals
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Synaptotagmin 1
- Antigensymbol:SYT1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Isotyp:IgG
- Reaktivitet:Rat
- Western blot:Yes
- Korsad adsorption:No
- Mått:Antibody fraction
- Form:Liquid
- Gen-ID:25716
- Antigen synonymer:P65
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Förvaringstemperatur:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-Synaptotagmin 1 Antibody was produced in rabbit by repeated immunizations with synthetic peptide corresponding to amino acid residues specific to synaptotagmin 1.
- Rening:affinity-purified
- Storlek:100 µl
- Förp.:100 µl
Specifikationer
Om denna produkt
Anti-Synaptotagmin 1 antibody is suitable for use in Western Blotting. Expect a band of approximately 65 kDa in size corresponding to synaptotagmin I Protein in the appropriate cell lysate or extract.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: SYT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat