Anti-SYN1 Rabbit Polyclonal Antibody
ROCK612-401-E33
- Antikroppstyp:Primär
- Antikroppsnamn:Synapsin 1
- Antigensymbol:SYN1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Rat
- Western blot:Yes
- ChIP:Yes
- Gen-ID:281510
- Storlek:100 µl
- Förp.:100 µl
Anti-Synapsin I pS62/pS67 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 78 kDa in size corresponding to synapsin I doublet phosphorylated at Ser62/67.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: SYN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat