Anti-DLG4 Rabbit Polyclonal Antibody
Katalog # ROCK612-401-E19
Leverantör: Rockland Immunochemicals
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Discs, large homolog 4 (Drosophila)
- Antigensymbol:DLG4
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Isotyp:IgG
- Reaktivitet:Rat
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:No
- Mått:Antibody fraction
- Form:Liquid
- Gen-ID:29495
- Antigen synonymer:Sap90|Dlgh4|PSD95
- Modifiering:phospho-Ser-95
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Förvaringstemperatur:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-PSD-95 Antibody was produced in rabbit by repeated immunizations with synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH.
- Rening:affinity-purified
- Storlek:100 µl
- Förp.:100 µl
Specifikationer
Om denna produkt
Anti-PSD 95 (Rabbit) antibody is suitable for use in Western Blotting. Expect a band of approximately 95 kDa in size corresponding to PSD-95 Protein in the appropriate cell lysate or extract.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: DLG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Rat