Anti-GST Mouse Monoclonal Antibody (DyLight® 649) [clone: 3D4]
Katalog # ROCK200-343-200
Leverantör: Rockland Immunochemicals
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Glutathione-S-Transferase
- Antigensymbol:GST
- Klonalitet:Monoclonal
- Klon:3D4
- Konjugation:DyLight® 649
- ELISA:Yes
- Gelskifte:Yes
- Värd:Mouse
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG1 kappa
- Reaktivitet:Multiple
- Western blot:Yes
- ChIP:Yes
- Neutralisation:Yes
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: GST
Clonality: Monoclonal
Clone: 3D4
Conjugation: DyLight® 649
Epitope:
Host: Mouse
Isotype:
Reactivity: pan