Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG (H+L) Goat Polyclonal Antibody (ATTO 655)
Anti-IgG (H+L) Goat Polyclonal Antibody (ATTO 655)
  ROCK610-157-121S
undefined
Anti-IgG (H+L) Goat Polyclonal Antibody (ATTO 655)
  ROCK610-157-121S

 

  • Antikroppstyp:
    Sekundär
  • Antigensymbol:
    IgG (H+L)
  • Klonalitet:
    Polyclonal
  • Konjugation:
    ATTO 655
  • Värd:
    Goat
  • ImmunoFluorescens:
    Yes
  • Reaktivitet:
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Mått:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Mouse IgG whole molecule
  • Tested applications:
    Fluoresence ELISA, Fluorescence Resonance Energy Transfer, Stimulated Emission Depletion Microscopy
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    100 µg
  • Förp.:
    100 µG

 

 

The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: ATTO 655
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse