Order Entry
Sweden
ContactUsLinkComponent
 
Anti-HIST2H3C Rabbit Polyclonal Antibody
Catalog # ROCK600-401-I72
undefined
Anti-HIST2H3C Rabbit Polyclonal Antibody
Catalog # ROCK600-401-I72

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    histone cluster 2, H3c
  • Antigensymbol:
    HIST2H3C
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Reaktivitet:
    Drosophila,
    Plant,
    Xenopus,
    Chicken,
    C. elegans,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • ChIP:
    Yes
  • Gen-ID:
    126961
  • Antigen synonymer:
    H3F2|H3.2|H3FM|H3|H3/M|H3FN
  • Modifiering:
    acetyl-Lys9
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,05% (w/v) Sodium Azide
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Histone H3 [ac Lys9] affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic acetylated peptide surrounding Lysine 9 of human Histone H3.2.
  • Tested applications:
    Dot Blot
  • Rening:
    purified by immunoaffinity chromatography
  • Storlek:
    50 µg
  • Förp.:
    50 µG

Specifications

About this item

Recommended Dilutions: ChIP: 2-5µg/million cells; Immunofluorescence: 1:100; Western Blot: 1:500; Immunochemistry: 1:100; Other Dilution: Dot Blot 1:1000

Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, C. elegans, Rat, Chicken, Xenopus, Drosophila, Plant