Order Entry
Sweden
ContactUsLinkComponent
Anti-HS3ST1 Goat Polyclonal Antibody
Anti-HS3ST1 Goat Polyclonal Antibody
Catalog # ORIGTA329076
Supplier:  OriGene
Anti-HS3ST1 Goat Polyclonal Antibody
Catalog # ORIGTA329076
Supplier:  OriGene

Specifications

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    HS3ST1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Goat
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Bovine
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Korsad adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA. Aliquot and store at -20°C. Minimize freezing and thawing.
  • Sequence:
    NKLHEYFHEPNKK
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    C Terminus (NKLHEYFHEPNKK)
  • Rening:
    purified by ammonium sulphate precipitation followed by antigen affinity chromatography
  • Storlek:
    100 μg
  • Förp.:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1-3?g/ml; ELISA: 1:128000

Type: Primary
Antigen: HS3ST1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Human