Order Entry
Sweden
ContactUsLinkComponent
Anti-HAPLN1 Rabbit Polyclonal Antibody
Anti-HAPLN1 Rabbit Polyclonal Antibody
Katalog # ORIGTA325115
Leverantör:  OriGene
Anti-HAPLN1 Rabbit Polyclonal Antibody
Katalog # ORIGTA325115
Leverantör:  OriGene

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Hyaluronan and proteoglycan link protein 1
  • Antigensymbol:
    HAPLN1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Flödescytometri:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    near N-terminus
  • Korsad adsorption:
    No
  • Form:
    Liquid
  • Aminosyretal:
    10 to 38
  • Storage buffer:
    Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
  • Molekylvikt:
    40.17 kDa
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    This HAPLN1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 10-38 amino acids from the N-terminal region of human HAPLN1.
  • Rening:
    purified by Protein A chromatography, followed by affinity purification
  • Storlek:
    400 μl
  • Förp.:
    400 µl

Specifikationer

Om denna produkt

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: HAPLN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human