Order Entry
Sweden
ContactUsLinkComponent
Anti-ATF2 Rabbit Polyclonal Antibody
Anti-ATF2 Rabbit Polyclonal Antibody
Katalog # ORIGTA324221
Leverantör:  OriGene
Anti-ATF2 Rabbit Polyclonal Antibody
Katalog # ORIGTA324221
Leverantör:  OriGene

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Activating transcription factor 240
  • Antigensymbol:
    ATF240
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Form:
    Liquid
  • Antigen synonymer:
    ATF2
  • Modifiering:
    phospho-Ser112 or 94
  • Storage buffer:
    PBS pH 7,3, 0,05% NaN3, 50% glycerol
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    PepT-ide sequence around phosphorylation site of serine 112 or 94 (D-L-S(p)-pL) derived from Human ATF2.
  • Rening:
    purified by antigen affinity chromatography
  • Storlek:
    100 μl
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

  • Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat