Order Entry
Sweden
ContactUsLinkComponent
Anti-IL18 Rabbit Polyclonal Antibody
Anti-IL18 Rabbit Polyclonal Antibody
Katalog # ORIGTA324190
Leverantör:  OriGene
Anti-IL18 Rabbit Polyclonal Antibody
Katalog # ORIGTA324190
Leverantör:  OriGene

Specifikationer

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    IL18
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    Full length
  • Korsad adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS pH 7,3, 0,05% NaN3, 50% glycerol
  • Molekylvikt:
    22 kDa
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length fusion protein
  • Rening:
    purified by antigen affinity chromatography
  • Storlek:
    100 μl
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

Recommended Dilutions: ELISA: 1:2000-5000; Western Blot: 1:1000-2000; Immunochemistry: 1:100-200

Type: Primary
Antigen: IL18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat