Order Entry
Sweden
ContactUsLinkComponent
Anti-GBP2 Rabbit Polyclonal Antibody
Anti-GBP2 Rabbit Polyclonal Antibody
Katalog # ORIGTA325034
Leverantör:  OriGene
Anti-GBP2 Rabbit Polyclonal Antibody
Katalog # ORIGTA325034
Leverantör:  OriGene

Specifikationer

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    GBP2
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    internal
  • Korsad adsorption:
    No
  • Form:
    Liquid
  • Aminosyretal:
    192 to 220
  • Storage buffer:
    Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
  • Molekylvikt:
    67.21 kDa
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    This GBP2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 192-220 amino acids from the Central region of human GBP2.
  • Rening:
    purified by Protein A chromatography, followed by affinity purification
  • Storlek:
    400 μl
  • Förp.:
    400 µl

Specifikationer

Om denna produkt

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: GBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human