Order Entry
Sweden
ContactUsLinkComponent
Anti-CDC37 Rabbit Polyclonal Antibody
Anti-CDC37 Rabbit Polyclonal Antibody
Catalog # ORIGTA322445
Supplier:  OriGene
Anti-CDC37 Rabbit Polyclonal Antibody
Catalog # ORIGTA322445
Supplier:  OriGene

Specifications

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    CDC37
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Form:
    Liquid
  • Aminosyretal:
    345 to 360
  • Storage buffer:
    PBS pH 7,3, 0,05% NaN3, 50% glycerol
  • Molekylvikt:
    44 kDa
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Synthetic peptide corresponding to a region derived from 345-360 amino acids of human cell division cycle 37 homolog (S. cerevisiae)
  • Rening:
    purified by antigen affinity chromatography
  • Storlek:
    100 μl
  • Förp.:
    100 µl

Specifications

About this item

Recommended Dilutions: ELISA: 1:1000-5000; Western Blot: 1:200-1000; Immunochemistry: 1:15-50

Type: Primary
Antigen: CDC37
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat