Anti-Ido1 Mouse Monoclonal Antibody [clone: 10.1]
Katalog # ORIGTA319542
Leverantör: OriGene
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Indoleamine 2,3-dioxygenase 1
- Antigensymbol:IDO1
- Klonalitet:Monoclonal
- Klon:10.1
- Konjugation:Unconjugated
- Värd:Mouse
- Reaktivitet:Human,Mouse
- Western blot:Yes
- Korsad adsorption:No
- Form:Liquid
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Förvaringstemperatur:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Anti-IDO1 (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with fragment of recombinant human and mouse IDO1 protein followed by hybridoma development.
- Storlek:100 μg
- Förp.:100 µG
Specifikationer
Om denna produkt
Recommended Dilutions: ELISA: 1:5000-1:50000; Western Blot: 1:500-1:1500; Immunochemistry: 1:100-1:500; Immunofluorescence: 1:50-1:100; Immunoprecipitation: 10-100 ?l
Type: Primary
Antigen: Ido1
Clonality: Monoclonal
Clone: 10.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse