Anti-S100A1 Rabbit Polyclonal Antibody
ORIGTA319193
: OriGene
- Antikroppstyp:Primär
- Antigensymbol:S100A1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Bovine
- Western blot:Yes
- Epitop:Full length
- Korsad adsorption:No
- Form:Liquid
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Förvaringstemperatur:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full-length bovine S100 protein (mixture of aa homodimers and ab heterodimers).
- Storlek:500 μg
- Förp.:500 µG
Recommended Dilutions: ELISA: 1:5000-1:25000; Western Blot: 1:500-1:3000; Immunochemistry: 1:200-1:2000
Type: Primary
Antigen: S100A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Bovine