Anti-KRT20 Mouse Monoclonal Antibody [clone: 6F6]
Catalog # ORIGCF802661
Supplier: OriGene
Specifications
- Antikroppstyp:Primär
- Antigensymbol:KRT20
- Klonalitet:Monoclonal
- Klon:6F6
- Konjugation:Unconjugated
- Värd:Mouse
- Isotyp:IgG2a
- Reaktivitet:Human
- Western blot:Yes
- Epitop:Full length
- Korsad adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molekylvikt:48.3 kDa
- Förvaringstemperatur:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human KRT20 (NP_061883) produced in E.coli.
- Rening:purified by affinity chromatography
- Storlek:100 μg
- Förp.:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: KRT20
Clonality: Monoclonal
Clone: 6F6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human