Order Entry
Sweden
ContactUsLinkComponent
Anti-UBE2B Mouse Monoclonal Antibody [clone: 1E11]
Anti-UBE2B Mouse Monoclonal Antibody [clone: 1E11]
Katalog # ORIGCF803573
Leverantör:  OriGene
Anti-UBE2B Mouse Monoclonal Antibody [clone: 1E11]
Katalog # ORIGCF803573
Leverantör:  OriGene

Specifikationer

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    UBE2B
  • Klonalitet:
    Monoclonal
  • Klon:
    1E11
  • Konjugation:
    Unconjugated
  • Värd:
    Mouse
  • Isotyp:
    IgG2b
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    Full length
  • Korsad adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molekylvikt:
    17.1 kDa
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human UBE2B (NP_003328) produced in E.coli.
  • Rening:
    purified by affinity chromatography
  • Storlek:
    100 μg
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: UBE2B
Clonality: Monoclonal
Clone: 1E11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human