Order Entry
Sweden
ContactUsLinkComponent
Anti-EPN2 Mouse Monoclonal Antibody [clone: 1C5]
Anti-EPN2 Mouse Monoclonal Antibody [clone: 1C5]
Catalog # ORIGCF504300
Supplier:  OriGene
Anti-EPN2 Mouse Monoclonal Antibody [clone: 1C5]
Catalog # ORIGCF504300
Supplier:  OriGene

Specifications

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    EPN2
  • Klonalitet:
    Monoclonal
  • Klon:
    1C5
  • Konjugation:
    Unconjugated
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG1
  • Reaktivitet:
    Monkey,
    Human
  • Western blot:
    Yes
  • Epitop:
    Full length
  • Korsad adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molekylvikt:
    68.3 kDa
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human EPN2(NP_055779) produced in HEK293T cell.
  • Rening:
    purified by affinity chromatography
  • Storlek:
    100 μg
  • Förp.:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: EPN2
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey