Anti-GALE Mouse Monoclonal Antibody [clone: 1C4]
Katalog # ORIGCF502146
Leverantör: OriGene
Specifikationer
- Antikroppstyp:Primär
- Antigensymbol:GALE
- Klonalitet:Monoclonal
- Klon:1C4
- Konjugation:Unconjugated
- Värd:Mouse
- ImmunKemi:Yes
- Isotyp:IgG1
- Reaktivitet:Monkey,Human,Dog,Rat
- Western blot:Yes
- Epitop:Full length
- Korsad adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molekylvikt:38.1 kDa
- Förvaringstemperatur:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human GALE (NP_000394) produced in HEK293T cell.
- Rening:purified by affinity chromatography
- Storlek:100 μg
- Förp.:100 µG
Specifikationer
Om denna produkt
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150
Type: Primary
Antigen: GALE
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat