Order Entry
Sweden
ContactUsLinkComponent
 
Anti-SNX8 Mouse Monoclonal Antibody [clone: 4B4]
  ORIGCF502134
 :  OriGene
Anti-SNX8 Mouse Monoclonal Antibody [clone: 4B4]
  ORIGCF502134
 :  OriGene

 

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    SNX8
  • Klonalitet:
    Monoclonal
  • Klon:
    4B4
  • Konjugation:
    Unconjugated
  • Flödescytometri:
    Yes
  • Värd:
    Mouse
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG2a
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    Full length
  • Korsad adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molekylvikt:
    52.4 kDa
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human SNX8 (NP_037453) produced in HEK293T cell.
  • Rening:
    purified by affinity chromatography
  • Storlek:
    100 μg
  • Förp.:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNX8
Clonality: Monoclonal
Clone: 4B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human