Order Entry
Sweden
ContactUsLinkComponent
Anti-PUS7 Mouse Monoclonal Antibody [clone: 4C6]
Anti-PUS7 Mouse Monoclonal Antibody [clone: 4C6]
  ORIGCF501816
 :  OriGene
Anti-PUS7 Mouse Monoclonal Antibody [clone: 4C6]
  ORIGCF501816
 :  OriGene

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Pseudouridylate synthase 7
  • Antigensymbol:
    PUS7
  • Klonalitet:
    Monoclonal
  • Klon:
    4C6
  • Konjugation:
    Unconjugated
  • Flödescytometri:
    Yes
  • Värd:
    Mouse
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG1
  • Reaktivitet:
    Monkey,
    Human,
    Dog,
    Rat
  • Western blot:
    Yes
  • Epitop:
    Full length
  • Korsad adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molekylvikt:
    74.9 kDa
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human PUS7 (NP_061915) produced in HEK293T cell.
  • Rening:
    purified by affinity chromatography
  • Storlek:
    100 μg
  • Förp.:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PUS7
Clonality: Monoclonal
Clone: 4C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat