Order Entry
Sweden
ContactUsLinkComponent
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 6E2]
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 6E2]
  ORIGCF501155
 :  OriGene
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 6E2]
  ORIGCF501155
 :  OriGene

 

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    CYP1A2
  • Klonalitet:
    Monoclonal
  • Klon:
    6E2
  • Konjugation:
    Unconjugated
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG2a
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Epitop:
    full length
  • Korsad adsorption:
    No
  • Form:
    Lyophilized
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molekylvikt:
    58.2 kDa
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length human recombinant protein of human CYP1A2 (NP_000752) produced in HEK293T cell.
  • Rening:
    purified by affinity chromatography
  • Storlek:
    100 μg
  • Förp.:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: CYP1A2
Clonality: Monoclonal
Clone: 6E2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human