Anti-TP53 Mouse Monoclonal Antibody [clone: 2E4]
Katalog # ORIGCF502783
Leverantör: OriGene
Specifikationer
- Antikroppstyp:Primär
- Antigensymbol:TP53
- Klonalitet:Monoclonal
- Klon:2E4
- Konjugation:Unconjugated
- Flödescytometri:Yes
- Värd:Mouse
- ImmunoFluorescens:Yes
- Isotyp:IgG1
- Reaktivitet:Monkey,Human,Mouse
- Western blot:Yes
- Epitop:Full length
- Korsad adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molekylvikt:43.5 kDa
- Förvaringstemperatur:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human TP53 (NP_000537) produced in HEK293T cell.
- Rening:purified by affinity chromatography
- Storlek:100 μg
- Förp.:100 µG
Specifikationer
Om denna produkt
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse