Order Entry
Sweden
ContactUsLinkComponent
Anti-LGALS7 Rabbit Polyclonal Antibody
Anti-LGALS7 Rabbit Polyclonal Antibody
Katalog # ORIGTA319111
Leverantör:  OriGene
Anti-LGALS7 Rabbit Polyclonal Antibody
Katalog # ORIGTA319111
Leverantör:  OriGene

Specifikationer

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    LGALS7
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    100 µg (0,5 mg/ml) affinity purified rabbit polyclonal antibody in phosphate-buffered saline (PBS) containing 30% glycerol, 0,5% BSA, and 0,01% thimerosal.
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Recombinant human Galectin-7
  • Rening:
    purified by affinity chromatography
  • Storlek:
    100 μg
  • Förp.:
    100 µG

Specifikationer

Om denna produkt

Recommended Dilutions: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: LGALS7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human