Order Entry
Sweden
ContactUsLinkComponent
Anti-DISC1 Rabbit Antibody
Anti-DISC1 Rabbit Antibody
Catalog # BSBTPA2023-1
Supplier:  Boster Bio
Anti-DISC1 Rabbit Antibody
Catalog # BSBTPA2023-1
Supplier:  Boster Bio

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Disrupted in schizophrenia 1
  • Antigensymbol:
    DISC1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonymer:
    SCZD9|KIAA0457|C1orf136
  • Aminosyretal:
    461 to 477
  • UniProtKB:
    Q9NRI5
  • Sequence:
    EKQQLQKEIEALQARMF
  • Förvaringstemperatur:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human DISC1 (461-477aa EKQQLQKEIEALQARMF), different from the related mouse sequence by two amino acids, and from the related rat sequence by four amino acids.
  • Rening:
    purified by immunogen affinity purification
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Disrupted in schizophrenia 1 protein(DISC1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

DISC1(Disrupted in Schizophrenia 1), is a protein that is encoded by the DISC1 gene in humans. Ma et al.(2002) determined that the mouse Disc1 gene maps to chromosome 8 in a region with homology of synteny to human chromosome 1q42. Ozeki et al.(2003) demonstrated that rodent Disc1 expression displayed pronounced developmental regulation, with the highest levels in late embryonic life during development of the cerebral cortex. Millar et al.(2005) showed that DISC1 interacts with the UCR2 domain of phosphodiesterase-4B, implicated in susceptibility to schizophrenia, and that elevation of cellular cAMP leads to dissociation of PDE4B from DISC1 and in increase in PDE4B activity.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: DISC1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse