Inverted microscope, Axio Vert.A1
Inverterad mikroskop, Binokular, 4× φ1, 10× φ1, 20× φ1, 40× φ1
Katalog # ZEIS4912370002000
Leverantör: ZEISS
Gå till:
Product Details & Documents
The Axio Vert.A1 is designed for the examination of heavy samples in biology.
- All contrast techniques available to see the finest structures
- Differential interference contrast (DIC) (optional)
- Circular polarised light: Differential interference contrast (C-DIC)
- Nosepiece turret (5×) can recognise a change of objectives
- Light manager to save and recover light intensity values
Leveransinformation: Supplied with an adapter and an object guide.
Specifikationer
- Förstoring:4× φ1, 10× φ1, 20× φ1, 40× φ1
- Beskrivning:Inverterad mikroskop
- Typ:Binokular
- Cat. No.:ZEIS4912370002000
- Belysning:Halogen
- Okular:WF 10×/23 mm
Accessories
Produktinformation
- Katalog #TillgänglighetPrice
- Transmitted light for LED illuminatorTransmitted light for LED illuminatorZEIS4230049020000Log in för att kolla tillgänglighet och ert pris1 * 1 Items
Produktinformation
- Katalog #TillgänglighetPrice
- Conversion filter, 5700 - 3200 K (Ø = 32 mm)Conversion filter, 5700 - 3200 K (Ø = 32 mm)ZEIS4278099000000Log in för att kolla tillgänglighet och ert pris1 * 1 Items