Nitrile gloves, Microflex® 93-843
Supplier: Ansell
Certificates
About this item
These disposable and durable gloves offer an advanced barrier protection against contaminants due to their lower acceptable pinhole rates (AQL 0,65).
- Non stick properties are built into the glove formulation to create a resistance to tape or adhesives, and to assist workers with improved efficiency and speed
- Contain no natural rubber latex or powder to help protect wearers from skin allergies, skin irritation and dryness
- Textured fingers ensure strong, efficient grip and excellent worker protection
- Polymer coating assures easy donning to help protect workers in fast-paced environments
Ideal for use in analytical testing, food processing, glass manufacturing, laboratory applications, patient examinations, light assembly tasks and pharmaceutical manufacturing.
Certifications: EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS; Cat. III; AQL 0,65
Food contact approved
Food contact approved
Specifications
- Catalog No:
- 112-4335
- 112-4336
- 112-4339
- 112-4338
- 112-4337
- Ej handspecifika/handspecifika:
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Yta:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- EN standard:
- EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS
- EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS
- EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS
- EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS
- EN 455, EN ISO 374-1:2016 Type B (JKPT), EN ISO 374-5:2016 VIRUS
- Godkända för livsmedelskontakt:
- +
- +
- +
- +
- +