Burner, Teclu
Catalog # RLAA4010
Supplier: R L ENTERPRISES
Specifications
- Beskrivning:Teclu burner
- Seq:01
- Förp.:1
- För:Coal gas
Specifications
About this item
The burner is constructed with a gas control needle valve. Specially designed conical mixing chamber is incorporated, which allows the burner to give off a very hot flame.
- Suitable for glass work and soldering
- Gas and air mixing is regulated by adjusting the knurled wheel below the tube
Height: 145 mm