PAR test agar
Katalog # LIOF11200
Leverantör: LIOFILCHEM
CAS-nummer:
Specifikationer
- Förp.:1
- Beskrivning:PAR test agar for antibiotic residue detection
Specifikationer
Om denna produkt
Medium with spores of Geobacillus stearothermophilus subsp. calidolactis for residual antibiotic detection in biological liquids.
- For milk, blood and urine