Culture media, in tubes
Catalog # HEIP1463030100
Supplier: MERCK LIFE SCIENCE
CAS Number:
Specifications
- Förp.:100
- Standard:EP, USP
- Beskrivning:Sabouraud dextrose agar
- Fyllning:18 ml
Specifications
About this item
Sabouraud dextrose is recommended for the microbial enumeration tests.