ContactUsLinkComponent
Round cuvettes
Round cuvettes
Catalog # HACHLYY621
Supplier:  Hach
undefined
Round cuvettes
Catalog # HACHLYY621
Supplier:  Hach
Specifications
  • Skiktdjup:
    11 mm
  • Volym:
    8 ml
  • Material:
    Glas
  • Beskrivning:
    Cuvette, for LICO
  • Förp.:
    560
Specifications
About this item
Round glass cuvette.