Flow meter
Flow meter, 10 - 100 l/hour, 210 mm
Katalog # BEHRB00217617
Leverantör: BEHR
Gå till:
Product Details & Documents
This adjustable flow through meter is a substitute of DFM for the determination of cyanide.
Specifikationer
- Flödeshastighet:10 - 100 l/hour
- Beskrivning:Flow meter
- Längd:210 mm