ContactUsLinkComponent
Calibration weights, OIML R111, class M2
Calibration weights, OIML R111, class M2
Katalog # ZWIEE201.12.11
Leverantör:  ZWIEBEL
Calibration weights, OIML R111, class M2
Katalog # ZWIEE201.12.11
Leverantör:  ZWIEBEL
Specifikationer
  • Vikt:
    1 kg
  • Förp.:
    1
  • Tolerans:
    160 ± mg
  • Typ:
    Class M2 weights with calibration certificate
Specifikationer
Om denna produkt
Weights made from brass. M2 weights are designed to calibrate masses of a lower accuracy class and be used with class II and III weighing instruments.
  • Ultra-compact and easy to handle
  • Weights are available as individual units or in sets (wooden box/base)