Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Media
Beredda mikrobiologiska media
Culture media, ready to use, in tubes
Culture media, ready to use, in tubes
Culture media, ready to use, in tubes
Catalog # HEIP1464290020
Leverantör:
MERCK LIFE SCIENCE
CAS-nummer:
Expand image
Culture media, ready to use, in tubes
Catalog # HEIP1464290020
Leverantör:
MERCK LIFE SCIENCE
CAS-nummer:
Specifikationer
Förp.:
20
Standard:
-
Beskrivning:
TSA tryptic soy agar
Seq:
06
Volym:
18 ml
Specifikationer
Om denna produkt
Product Details
Documentation
Product Details
Documentation