ContactUsLinkComponent
Water treatment units: Acqua purificator G 7895/1
Water treatment units: Acqua purificator G 7895/1
Catalog # 141-1230
Supplier:  MIELE
undefined
Water treatment units: Acqua purificator G 7895/1
Catalog # 141-1230
Supplier:  MIELE
Specifications
  • Beskrivning:
    Water purifier, Aqua purificator, G 7895/1, front loading cabinet, stainless steel casing, angled fascia, freestanding, can be built under the plan, for use with E 310 − E318 cartridges, for G 78XX series
  • Seq:
    02
  • Förp.:
    1
Specifications
About this item
Water treatment units for use with G 78XX series machines. Cabinet for two E310/318 water demineralisation cartridges with conductivity meter, generally recommended quality for final rinse <15 µs/cm.
  • Two pressure hoses, approx. 1,2 m long, with ³/₄" threated union
  • 1× cold water connection to cartridge, 2,5 to 10 bar pressure
  • 1× connection from cartridge to machine (loss of pressure approx. 1 bar for each cartridge)
Casing made from white steel or stainless steel.