Detergent, Disinfectant, Oxivir® Plus
Leverantör: Diversey
Certifikat
Om denna produkt
Cleaner disinfectant that eliminates a broad spectrum of microorganisms (bacteria, viruses, mold fungi, yeasts). It utilises the environmentally friendly Accelerated Hydrogen Peroxide (AHP®) technology: When using the product it decomposes into water and oxygen.
- Safe on all acid resistant surfaces including upholstery and carpets
- Ideal for prevention of outbreaks and as part of a body spillage programme
- Woolsafe approved for soft furnishings
Certifications: EN 14476 Norovirus in 2 minutes. EN 1276 E. Coli in 5 minutes. EN 1650 in 15 minutes.
Caution: Use biocides safely. Always read the label and product information before use.